Skip to Content
Merck
All Photos(2)

Key Documents

810151P

Avanti

14:0-06:0 NBD PE

Avanti Research - A Croda Brand 810151P, powder

Synonym(s):

1-myristoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H52N5O11P
CAS Number:
Molecular Weight:
701.75
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810151P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810151P

shipped in

dry ice

storage temp.

−20°C

General description

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) is a fluorescently labeled lipid localized at the membrane interface. The molecule contains fluorescent NBD label attached to the headgroup of phosphatidylethanolamine. Phosphoethanolamine is the second most abundant, metabolically related aminophospholipid, mostly enriched in brain.

Biochem/physiol Actions

Phosphoethanolamine of cardiac myocytes minimizes the cell damage after ischemia. N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) aids is monitoring the Red edge excitation shift (REES) effects with the unique motional and dielectric properties it possesses.

Packaging

5 mL Amber Glass Screw Cap Vial (810151P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of NBD-labeled lipids in membrane and cell biology
Fluorescent methods to study biological membranes, 37-50 (2012)
S C Lopes et al.
Biochimica et biophysica acta. Biomembranes, 1861(6), 1152-1161 (2019-03-07)
Mitochondrial membranes are pointed out as the site of cardiotoxic action of local anaesthetics. Its three main phospholipids components are phosphatidylcholine, phosphatidylethanolamine and cardiolipin. Cardiolipins, in eukaryotes, are only found in mitochondria and are essential for the maintenance of its
Jean E Vance et al.
Biochimica et biophysica acta, 1831(3), 543-554 (2012-09-11)
Phosphatidylserine (PS) and phosphatidylethanolamine (PE) are metabolically related membrane aminophospholipids. In mammalian cells, PS is required for targeting and function of several intracellular signaling proteins. Moreover, PS is asymmetrically distributed in the plasma membrane. Although PS is highly enriched in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service