Skip to Content
Merck
All Photos(1)

Key Documents

790528C

Avanti

18:1 DGS-NTA

1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (ammonium salt), chloroform

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (ammonium salt); DOGS NTA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H100N5O13
CAS Number:
Molecular Weight:
1015.39
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (790528C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790528C

concentration

5 mg/mL (790528C-5mg)

shipped in

dry ice

storage temp.

−20°C

Application

18:1 DGS-NTA has been used for the preparation of liposomes.

Packaging

5 mL Clear Glass Sealed Ampule (790528C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Actin retrograde flow and actomyosin II arc contraction drive receptor cluster dynamics at the immunological synapse in Jurkat T cells
Yi J, et al.
Molecular Biology of the Cell, 23(5), 834-852 (2012)
E Barklis et al.
The Journal of biological chemistry, 273(13), 7177-7180 (1998-04-29)
In an in vitro system that mimics the assembly of immature human immunodeficiency virus (HIV) particles, ordered arrays of HIV-1 capsid (CA) proteins encoded by the viral gag gene have been obtained by incubation of histidine-tagged capsid proteins (His-HIVCA) beneath
C Dietrich et al.
Biochemistry, 35(4), 1100-1105 (1996-01-30)
The coupling of a DNA-binding protein to self-organized lipid monolayers is examined at the air-water interface by means of film balance techniques and epifluorescence microscopy. We used two recombinant species of the heat shock factor HSF24 which differ only in
E Barklis et al.
The EMBO journal, 16(6), 1199-1213 (1997-03-17)
We have developed a system for analysis of histidine-tagged (His-tagged) retrovirus core (Gag) proteins, assembled in vitro on lipid monolayers consisting of egg phosphatidylcholine (PC) plus the novel lipid DHGN. DHGN was shown to chelate nickel by atomic absorption spectrometry
C Vénien-Bryan et al.
Journal of molecular biology, 274(5), 687-692 (1998-02-07)
Two-dimensional crystals of the histidine-tagged-HupR protein, a transcriptional regulator from the photosynthetic bacterium Rhodobacter capsulatus, were obtained upon specific interaction with a Ni2+-chelated lipid monolayer. HupR is a response regulator of the NtrC family; it activates the transcription of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service