Merck
Organization of HIV-1 capsid proteins on a lipid monolayer.
The Journal of biological chemistry (1998-04-29)
E Barklis, J McDermott, S Wilkens, S Fuller, D Thompson
ABSTRACT
In an in vitro system that mimics the assembly of immature human immunodeficiency virus (HIV) particles, ordered arrays of HIV-1 capsid (CA) proteins encoded by the viral gag gene have been obtained by incubation of histidine-tagged capsid proteins (His-HIVCA) beneath lipid monolayers containing the nickel-chelating lipid, 1,2-di-O-hexadecyl-sn-glycero-3-(1'-2"-R-hydroxy-3'-N-(5-amino-1- carboxypentyl)iminodiacetic acid)propyl ether. The membrane-bound His-HIVCA proteins formed small crystalline arrays of primitive (p1) unit cells with dimensions of a = 74.2 A, b = 126.2 A, gamma = 89.3 degrees. The image-analyzed two-dimensional projection of His-HIVCA assemblies shows a cage-like lattice, consisting of hexamer and trimer units, surrounding protein-free cage holes. The hexamer-coordinated cage holes of 26.3-A diameter are spaced at 74. 2-A intervals: these distances, and the hexamer-trimer arrangement, are consistent with previous, lower resolution studies on immature HIV-1 virus particles produced in vivo. Additionally, HIV-1 matrix protein trimer unit structures align to the His-HIVCA trimer units such that residues previously shown to interact with the HIV-1 gp120/gp41 envelope protein complex are oriented toward the hexamer cage holes. Our results form a bridge between results from conventional methods for the analysis of HIV particle structure.
Product Description
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Avanti
18:1 DGS-NTA, 1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (ammonium salt), chloroform
Avanti
18:1 DGS-NTA, 1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (ammonium salt), powder
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand