Skip to Content
Merck
All Photos(1)

Key Documents

181104C

Avanti

Liver Extract Total

Avanti Research - A Croda Brand

Synonym(s):

Liver Total Lipid Extract (Bovine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

chloroform solution

packaging

pkg of 1 × 4 mL (181104C-100mg)
pkg of 5 × 4 mL (181104C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

25 mg/mL (181104C-100mg)
25 mg/mL (181104C-500mg)

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Total Lipid Extract is a chloroform:methanol extract of the respective tissue (Brain, Soybean, Heart, etc.). This extract is partitioned against deionized water, and the chloroform phase is concentrated.

Application

Liver extract total has been used for the identification of cholesterol as an endogenous ligand of human macrophage-inducible C-type lectin (hMincle).

Packaging

5 mL Clear Glass Sealed Ampule (181104C-100mg)
5 mL Clear Glass Sealed Ampule (181104C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Human Mincle binds to cholesterol crystals and triggers innate immune responses
Kiyotake R, et al.
The Journal of biological chemistry, 290(42), 25322-25332 (2015)
Hari Kiran Kotapati et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1145, 122099-122099 (2020-04-20)
Three normal phase HPLC methods were produced to separate lipid classes on a PVA-Sil stationary phase including: 9 polar lipids (method 1); 13 combined polar and neutral lipids (method 2); and a combined method that further separates the neutral lipids
Ryoko Kiyotake et al.
The Journal of biological chemistry, 290(42), 25322-25332 (2015-08-25)
C-type lectin receptors (CLRs) are an emerging family of pattern recognition receptors that recognizes pathogens or damaged tissue to trigger innate immune responses. However, endogenous ligands for CLRs are not fully understood. In this study, we sought to identify an
Filip Duša et al.
Biochimica et biophysica acta. Biomembranes, 1862(2), 183115-183115 (2019-11-11)
The cell membrane is mainly composed of lipid bilayers with inserted proteins and carbohydrates. Lipid bilayers made of purified or synthetic lipids are widely used for estimating the effect of target compounds on cell membranes. However, the composition of such
Giuseppe Paglia et al.
Nature protocols, 12(4), 797-813 (2017-03-17)
Metabolomics and lipidomics aim to profile the wide range of metabolites and lipids that are present in biological samples. Recently, ion mobility spectrometry (IMS) has been used to support metabolomics and lipidomics applications to facilitate the separation and the identification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service