Skip to Content
Merck
All Photos(1)

Key Documents

Z742576

Adapter pack for MyFuge 5 Microcentrifuge, holds 0.5-2.0 mL cryovials and 1.5/2.0 mL HPLC vials

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.78

feature

Holds 0.5 to 2.0 mL cryotubes tubes

packaging

pack of 4 ea

manufacturer/tradename

Benchmark Scientific C1005-AC2

technique(s)

centrifugation: suitable

compatibility

for use with MyFuge 5 Microcentrifuge

General description

Benchmark Scientific Adapter pack, for Cryovials (0.5 to 2.0ml) and 1.5/2.0ml HPLC vials

Legal Information

MyFuge is a trademark of Benchmark Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service