CLS432057
Corning® CoolRack®
96 x 1 mL, capacity 96 x 1.4 mL 2D tubes
Synonym(s):
Staining, Staining Accessories
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122806
NACRES:
NB.12
Recommended Products
material
foamed PE
feature
wells 96
Holds 96 x 1 mL tubes
packaging
pkg of 1
manufacturer/tradename
Corning 432057
technique(s)
cryopreservation: suitable
L × W × H
13.2 cm × 8.9 cm × 4.3 cm
size
14 mm , row spacing
17.8 mm , column spacing
well depth
14 mm
well diam.
11.1 mm
Looking for similar products? Visit Product Comparison Guide
General description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
- Ensures +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
- Certain CoolRack modules are SBS-compatible.
Legal Information
CoolRack is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service