Z742017
Duran® Youtility Pouring RingGL 45
cyan, polypropylene (food grade)
Synonym(s):
Duran® Youtility
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
polypropylene (food grade)
packaging
pkg of 16 ea
manufacturer/tradename
DURAN® 292412808
color
, cyan
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Specially shaped gripping zones on both sides of the bottle for easier and safer handling
- Opening or closing of the bottle is faster, and fully compatible with DIN GL 45 closures and other accessories
- Slimmer YOUTILITY bottle shape allows a more optimal use of limited space in autoclaves and laboratory refrigerators
- Pre-defined labelling area is compatible with DURAN self-adhesive YOUTILITY bottle labels
- Nominal volume is shown at the top of the easy-to-read graduation scale for fast determination of the volumes
- Sequentially sized bottles are available in four volumes
- Moulded from the DURAN borosilicate 3.3 pharmacopeia type 1 neutral glass
- Retrace Code for full product traceability
- Each bottle is supplied as a complete system, with a pouring ring (cyan polypropylene) and a GL 45 cap (cyan polypropylene)
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service