Key Documents
Z680397
Duran® GL 45 connection system caps and accessories
GL 45 blue PP screw cap with 2 x GL 14 ports
Synonym(s):
1129750, Duran Cap Accessories, GL 45 Connection Caps
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Pricing and availability is not currently available.
material
blue-gray polypropylene
manufacturer/tradename
DURAN® 1129750
parameter
-40-140 °C temp. limit
General description
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–2 of 2 Questions  
  1. Hi, What bottle would be best to use with cap GL 45 Z680397 with 6 mm diameter ports? I am looking to use a bottle of 250 to 500 ml. Thank you
    1 answer
    1. These GL 45 connection system caps are suitable for all bottles with a GL 45 thread. Please see the links below to review several suitable bottle options available:
      https://www.sigmaaldrich.com/product/aldrich/z742009
      https://www.sigmaaldrich.com/product/aldrich/z742012
      https://www.sigmaaldrich.com/product/aldrich/z742008
      Helpful?
  2. Hi, I am wondering if all the following parts are included in your product. Insert for screw cap GL 14, 3.0 mm (2 pcs per one cap) and Screw Cap GL 14 Hose Connection (2 pcs per one cap). Thanks.
    1 answer
    1. This product is a GL45 screw cap with two ports that have GL14 threads as shown in product image. The inserts and hose connections are not included, but are sold separately.
      Helpful?