Key Documents
Z569828
2mag MIXcontrol 20 control unit for MIXdrive
AC/DC input 100 - 240 V AC, Schuko plug
Synonym(s):
control unit, power bank, power unit
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
feature
CE compliant
packaging
pack of 1 ea
manufacturer/tradename
2mag 90200
Features and Benefits
  • Control unit with 20 watts power
  • Speed range from 100 up to 1,600 rpm
  • Jerk-free stirring also at low speeds
  • Power setting (4-steps) for high power with regard to large and viscous stirring amounts and reduced power for the warming-free continuous operation of MIXdrive stirring drives
  • Clear digital display for speed- and power settings
  • SoftStart for reliable catching/centering and safe accelerating of the stirring bar
  • QuickSet for quick setting of start- and maximum speed, automatic memory function to save the last settings, resistant stainless steel housing, easy to clean
Choose from one of the most recent versions:
Certificates of Analysis (COA)