Key Documents
Z569666
KDS syringe multi-rack for Legato series pumps
microliter
Synonym(s):
dosing pump syringe rack, infusion pump syringe rack
About This Item
UNSPSC Code:
41122800
NACRES:
NB.01
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
KD Scientific 78-8302
General description
Accommodates up to 4 x 0.5 μL to 10 mL syringes. Works with Legato 200, 210 and 210P.
Legal Information
Legato is a trademark of KD Scientific
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–2 of 2 Questions  
  1. What is the size of the syringes that can be used with this device.
    1 answer
    1. As mentioned in the 'PROPERTIES' section, this product accommodates 0.5 ?L to 10 mL syringes.
      Helpful?
  2. What is the slowest possible flow rate? 
    1 answer
    1. This product is distributed on behalf of KD Scientific. This is the Microliter Syringe Multi-Rack to the pump. The manufacturer states the flow range for the KDS Legato™ 200 series syringe pump can be controlled from 5 pl/min to 215.803 ml/min. The syringe pump is available at this link: https://www.sigmaaldrich.com/US/en/product/aldrich/z569585
      Helpful?