Key Documents
Z562505
LabJaws swivel clamp holder
jaw capacity 0-19 mm (0-0.75 in.), nickel plated (zinc)
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
material
nickel plated (zinc)
manufacturer/tradename
Troemner 916065
jaw capacity
0-19 mm (0-0.75 in.)
General description
Corrosion resistant. Has precise fit and powerful holding ability. Designed to enable tilting of clamp at any angle in parallel planes. Outside adjustment screw permits close distance between items being held.
Legal Information
LabJaws is a trademark of Henry Troemner LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)