Key Documents
Z557579
Sigma-Aldrich
Aldrich® pear-shaped splash-guard adapter
capacity 100 mL, female joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
female joint: ST/NS24/40
male joint ST/NS14/20
description
anti-climb
capacity
100 mL
Related Categories
General description
Application: for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of "bumping. Tapered sidewalls eliminate hold up common to round style adapters and permits continuous washing with condensed solvent.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)