Key Documents
Z557420
Julabo® bath spring clamps for Erlenmeyer flasks
fits 25 mL flasks
About This Item
UNSPSC Code:
41122413
NACRES:
NB.77
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970602
General description
The individual spring clips can be positioned in the shaking water bath using the base tray for spring clips (Z743201-1EA). A maximum of 28 spring clips for 25 ml Erlenmeyer flasks can be mounted on a base tray.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)