Key Documents
Z256803
Ace Allihn condenser with Ace-Thred hose connections
joint: ST/NS 24/40, jacket L 300 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint: ST/NS24/40
manufacturer/tradename
Ace Glass 5946118
jacket L
300 mm
Related Categories
General description
Fitted with #7 Ace-Thred hose connections for 0.25 in. I.D. tubing. Features threaded inlet/outlet connections that incorporate UHDPE (ultra-high-density PE) connectors and nylon bushings to make easy connect/disconnect of coolant tubing. The bushing compresses the connector and a silicone O-ring for a leaktight seal. Approximately one bulb per 50mm.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)