Key Documents
Z551031
Graduated Valumetric bottles
clear soda-lime glass bottle, capacity 250 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 24 ea
Synonym(s):
W216974
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Pricing and availability is not currently available.
material
(PTFE faced LDPE liner)
clear soda-lime glass bottle
round bottle
white polypropylene cap
Joint
Joints threaded neck (58-400)
feature
with cap
packaging
case of 24 ea
manufacturer/tradename
WHEATON® W216974
capacity
250 mL
diam. × H
62 mm × 127 mm
General description
Ideal for use as sample bottles. They are also useful for dry powders, soil samples, and specimen storage. All bottles are made of clear, Type III soda lime glass.
Legal Information
Valumetric is a trademark of Wheaton Industries, Inc.
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)