Z530603
Dropping bottle with graduated dropper
capacity 30 mL
Synonym(s):
dropper bottle, dropper bottles
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
colorless bottle
glass bottle
round bottle
Joint
joint ST/NS neck14/20
packaging
pack of 1 ea
capacity
30 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Clear glass dropping bottle with ground glass joint. The 1 mL dropper has 0.10 mL graduations.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service