Z403571
IKA® supports for stirring hot plates
H 44 boss head clamp, cast aluminum, 1/cs
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.76
Recommended Products
material
aluminum (cast aluminum)
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
IKA 0002437700
color
none
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
For fastening the holding rod H 38 to the support rod H 16 V.
Clamping range - stand: 10 - 11 mm
Clamping range - extension arm: 11 mm
Material: cast aluminium
Clamping range - stand: 10 - 11 mm
Clamping range - extension arm: 11 mm
Material: cast aluminium
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service