Skip to Content
Merck
All Photos(1)

Key Documents

Z341487

Sigma-Aldrich

Hirschmann® Ceramus® bottle-top dispenser, adjustable volume

dispensing volume 1.0-5.0 mL, precision: 0.1 CV%

Synonym(s):

Adjustable liquid dispenser, Bottle-top dispenser, Precision dispenser

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

subdivision 0.1 mL
with recirculating valve

manufacturer/tradename

Hirschmann 933200010

dispensing volume measuring range

1.0-5.0 mL, precision: 0.1 CV%

volume

1-5 mL

fitting

GL 32
GL 38
GL 45
S 40

Looking for similar products? Visit Product Comparison Guide

General description

  • Chemically inert − unique ceramic dispenser piston in a Duran® borosilicate glass barrel
  • Micro-step volume adjustment − ensures reproducibility
  • Rotates on the bottle − for convenience
  • Recirculation system − for fast priming without loss of reagent
  • Lifetime calibration − supplied with individual serial number and Test Certificate

Includes adapters for:

  • GL 45 threads/Pyrex® and Duran® media bottles
  • GL 32 threads/Aldrich Sealed for Quality and Sure/Seal bottles
  • GL 38 threads/Gallon bottles
  • S40 threads

Specifications:

  • Piston − 99.7% pure Al2O3
  • Barrel − Duran® borosilicate glass
  • Valve block and housing − ECTFE
  • Valves − borosilicate glass ball and seat (HF: ceramic); Hastelloy® discharge valve spring
  • Suction and discharge tubes − FEP
  • Sterilization − Steam sterilizable at 121°C/2 bar, complete unit without disassembly
  • Luer-lock connection − for filters and drying tubes

Legal Information

Ceramus is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
DURAN is a registered trademark of DWK Life Sciences
Hastelloy is a registered trademark of Haynes International, Inc.
Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service