Key Documents
Z258997
Flexible stirrer shaft
L 152.4 cm (60 in.) , overall
About This Item
UNSPSC Code:
41103800
NACRES:
NB.72
material
glass
packaging
pack of 1 ea
manufacturer/tradename
Ace Glass 8081-32
L
152.4 cm (60 in.) , overall
General description
Used for isolating motor from stirring shaft. Designed with a ball bearing motor coupling at one end for connection to any motor with an 8 mm (5/16 in.) diam. shaft or other power source using an 8 mm diam. round steel adapter (included).
The other end has a detachable handpiece with an 8 mm diam. round steel adapter for connection to a FLEX-GRIP® stirrer chuck (available separately). Handpiece is secured with a lab clamp. Shafts operate to 14,000 rpm in a counter-clockwise direction.
Operating and service instructions for lubrication are supplied.
Complete item includes: A flexible shaft, motor coupling for 8 mm shaft, handpiece with adapter for chuck, and a chuck wrench and chain.
Legal Information
Flex-Grip is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)