Key Documents
Z257419
Spirit-filled low-temperature thermometer
L 305 mm (30.5 cm), temp. range -200/30 °C
About This Item
UNSPSC Code:
41112303
NACRES:
NB.79
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B60760-1600
parameter
-200/30 °C temp. range
L
305 mm (30.5 cm)
General description
These thermometers contain an organic liquid except Z257419 which is filled with a clear Toluene liquid. Total-immersion. 1° divisions for use in general-laboratory, low-temperature applications.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)