Key Documents
Z203610
Sigma-Aldrich
Black plastic caps
Solid-top, PTFE-faced rubber liner, thread for 15-415
Synonym(s):
240463
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
packaging
pkg of 200 ea
manufacturer/tradename
WHEATON®
fitting
thread for 15-415
General description
These autoclavable caps are designed to ensure the ultimate in product safety.
Features and Benefits
• PTFE faced (14B) styrene-butadiene rubber liner provides a totally inert inner seal and surface facing the sample or product•
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)