Key Documents
WHAWB120028
QIAcard FTA CloneSaver Cards
96 well format, pkg of 5 cards
Synonym(s):
QIAcard FTA CloneSaver (5), Whatman FTA, Z747440
About This Item
UNSPSC Code:
41105500
NACRES:
NB.22
description
CloneSaver Card, 96 well format, 5 μL maximum volume/sample area, 480 μL maximum total volume/card
manufacturer/tradename
Qiagen WB120028
packaging
pkg of 5 cards
Related Categories
General description
CloneSaver Card is designed for the collection, long-term storage, and purification of plasmid and BAC DNA from bacterial clones in a 96-well format
Features and Benefits
  • Captures and stabilizes bacterial artificial chromosome (BAC) and plasmid DNA in one easy step: without the requirement of reagents, centrifuges or other materials
  • Serves as a secure, reliable alternative to glycerol stocks
  • Simplifies storage, transport and access to clone libraries: conveniently, easily and inexpensively
  • Ensures faster clone identification and recovery through sample ID chart, instruction for use and area for bar code labeling
  • Produces ready-to-use vector DNA in less than 30 minutes: polymerase chain reaction (PCR) ready bacterial clones
  • Protects clone stocks from phage infections by inactivating bacteria and phage
Other Notes
Field of Use : For internal research use only
Legal Information
FTA is a trademark of Qiagen Group
QIAcard is a trademark of Qiagen Group
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Description of common problems that can occur during genomic DNA preparation
This article describes the evaluation of Whatman FTA cards from Cytiva for their ability to collect, store, and isolate high-quality RNA from a variety of crude biological samples.
Polymerase chain reaction (PCR) is a technique that results in exponential amplification of a target DNA sequence.
Protocols
This study examined the yield and quality of DNA from samples applied to Whatman FTA cards, using five common methods of DNA extraction.
A number of options are available for sample collection, transport, archiving, and DNA purification.
A number of options are available for sample collection, transport, archiving, and DNA purification.
A number of options are available for sample collection, transport, archiving, and DNA purification.
See All