Key Documents
WHAWB120401
QIAcard FTA Elute Micro cards
non-indicating, 4 sample areas per card, pkg of 25 cards
Synonym(s):
QIAcard FTA Elute Micro (25), Whatman FTA, Z747556
About This Item
UNSPSC Code:
41105500
NACRES:
NB.22
manufacturer/tradename
Qiagen WB120401
packaging
pkg of 25 cards
General description
FTA Cards offer a compact room-temperature storage system that reduces the need for precious freezer space. Unlike the QIAcard FTA cards, the FTA Elute cards feature a technology for convenient water based elution of nucleic acids.
QIAcard FTA Elute Micro format with 4 spot area (spot volume 12-40 µL) to collect, stabilize, process, transport, and archive colored samples such as blood. FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Convenient for protocols that require different locations for testing and archiving samples. Different samples can be processed independently.
Features and Benefits
  • Two sample areas for application of up to 250 μl sample volume per card.
  • Simply apply your sample to the FTA Card. Cell membranes and organelles are lysed and the released nucleic acids are entrapped in the fibers of the matrix. The nucleic acids remain immobilized and are preserved for transport, immediate processing or long-term room temperature storage.
  • Since captured nucleic acids are preserved, FTA Cards facilitate sample collection in remote locations and simplify sample transport.
  • Ship your samples back to the laboratory without expensive special handling or dry ice and process at your convenience.
  • FTA Cards can be used with virtually any sample type: Blood, Cultured cells, Buccal cells, Plasmids, and Solid tissue.
  • Captured nucleic acids are ready for purification when you are. The FTA Elute technology enables convenient water-based elution of nucleic acids for downstream experiments.
Other Notes
Field of Use : For internal research use only
Legal Information
FTA is a trademark of Qiagen Group
QIAcard is a trademark of Qiagen Group
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Articles
This page shows downstream applications after genomic DNA purification.
Description of common problems that can occur during genomic DNA preparation
Polymerase chain reaction (PCR) is a technique that results in exponential amplification of a target DNA sequence.
Protocols
This page shows genomic DNA purification from sample on FTA elute from Cytiva.
A number of options are available for sample collection, transport, archiving, and DNA purification.
A number of options are available for sample collection, transport, archiving, and DNA purification.
A number of options are available for sample collection, transport, archiving, and DNA purification.
See All