Skip to Content
Merck
All Photos(1)

Key Documents

WHA1822025

Whatman® glass microfiber grade GF/C filter discs 1.2 μm pore size

white, binder free, 0.26 mm thick, 100 ea, 25 mm diam

Synonym(s):

Z242349, glass fiber filters, glass microfiber filters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
NACRES:
NB.24

product name

Whatman® glass microfiber filters, binder free, Grade GF/C, Grade GF/C circles, 25 mm, 100/pk

material

borosilicate glass
circles

description

GF/C grade: medium to fast, high loading

sterility

non-sterile

packaging

pack of 100

manufacturer/tradename

Whatman 1822-025
Whatman Article No. 28418377 (US reference)

diam.

25 mm

thickness

0.26 mm

pore size

1.2 μm

suitability

suitable for (collection of suspended solids in potable water and natural and industrial wastes)

Looking for similar products? Visit Product Comparison Guide

General description

Whatman® Filter Paper grade GF/C glass microfiber filters provide good loading capacity. GF/C grade filter papers support the analysis of total suspended solids.
Grade GF/C: 1.2 μm


Cytiva’ Whatman Filter Paper grade GF/C glass filters offer fine particle retention, good flow rate and loading capacity. GF/C filter papers support total suspended solids analysis.

Application

Widely used for cell harvesting, liquid scintillation counting and binding assays, where more loading capacity is required. Highly suitable for use in applications involving air filtration and for the gravimetric analysis of volatile materials, where ignition is involved.

Features and Benefits

  • Widely used as a total suspended solids filter for applications testing both potable water and waste water.

  • Widely used as a total suspended solids filter for applications testing both potable water and waste water.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service