Key Documents
WHA10534321
Whatman® protein saver card accessories
Foil barrier reusable bags, pack of 100 ea
Synonym(s):
Z699527, foil bag, foil bags
About This Item
UNSPSC Code:
41111700
NACRES:
NB.79
packaging
pack of 100 ea
manufacturer/tradename
Whatman 10534321
Whatman Article No. 28416623 (US reference)
General description
These bags effectively safeguard contents from moisture and contaminants while ensuring easy access.
Features and Benefits
  • Foil bag with size 3.875” × 5.6875” and zipper to close.
  • Suitable for use with Whatman 903 cards.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)