W396400
2-Acetyl-3-methylpyrazine
≥98%, FG
Synonym(s):
1-(3-methylpyrazin-2-yl)ethanone
About This Item
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Fragrance grade
Halal
Kosher
Agency
follows IFRA guidelines
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
Assay
≥98%
refractive index
n20/D 1.521 (lit.)
bp
90 °C/20 mmHg (lit.)
density
1.114 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
Organoleptic
nutty; roasted
storage temp.
2-8°C
SMILES string
CC(=O)c1nccnc1C
InChI
1S/C7H8N2O/c1-5-7(6(2)10)9-4-3-8-5/h3-4H,1-2H3
InChI key
QUNOTZOHYZZWKQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- RIFM fragrance ingredient safety assessment, 2-acetyl-3-methylpyrazine, CAS Registry number 23787-80-6.: This study, published in "Food and Chemical Toxicology," assesses the safety of 2-Acetyl-3-methylpyrazine as a fragrance ingredient. The comprehensive evaluation covers its toxicological profile, exposure levels, and safety margins to ensure it is safe for use in consumer products (Api et al., 2023).
- New propositions about coffee wet processing: Chemical and sensory perspectives.: Published in "Food Chemistry," this paper investigates the effects of different wet processing methods on the chemical composition and sensory attributes of coffee, highlighting the role of 2-Acetyl-3-methylpyrazine in flavor development (Pereira et al., 2020).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
176.0 °F - closed cup
Flash Point(C)
80 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service