Skip to Content
Merck
All Photos(2)

Key Documents

W353205

Sigma-Aldrich

3-Decen-2-one

predominantly trans, ≥97%, stabilized, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH=CHCOCH3
CAS Number:
Molecular Weight:
154.25
FEMA Number:
3532
EC Number:
Council of Europe no.:
11751
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.121
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥97%

contains

α-Tocopherol. synthetic as stabilizer

refractive index

n20/D 1.45 (lit.)

bp

125 °C/12 mmHg (lit.)

density

0.847 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; green; jasmine; vegetable

SMILES string

CCCCCC\C=C\C(C)=O

InChI

1S/C10H18O/c1-3-4-5-6-7-8-9-10(2)11/h8-9H,3-7H2,1-2H3/b9-8+

InChI key

JRPDANVNRUIUAB-CMDGGOBGSA-N

General description

3-Decen-2-one occurs naturally in certain species of mushroom.

Application

Possible applications: Apricot, peach, watermelon, pear, cucumber, and tropical fruits, dairy flavors such as milks creams and cheeses. Fatty notes in chicken, fat replacers and lard. Used in mandarin, tangerine, grapefruit and other citrus flavors.

Biochem/physiol Actions

Taste at 1 to 5 ppm

Other Notes

Natural occurrence: Mushrooms.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Volatile Composition of Some Cultivated and Wild Culinary-Medicinal Mushrooms from Hungary.
Csoka M, et al.
International Journal of Medicinal Mushrooms, 19(5) (2017)
Svenja Nörenberg et al.
Journal of agricultural and food chemistry, 65(40), 8913-8922 (2017-09-19)
A homologous series of 4-mercapto-2-alkanols (C5-C10) was used to investigate the impact of the stereochemistry on the sensory properties of a class of naturally occurring polyfunctional thiols having a 1,3-oxygen-sulfur functionality. Stereoisomers were obtained via syntheses of racemic mixtures and
Character impact odorants from wild mushroom (Lactarius hatsudake) used in Japanese traditional food.
Miyazawa M, et al.
Flavour and Fragrance Journal, 25(4), 197-201 (2010)
Mosciano, G.
Perfumer & Flavorist, 25, 71-71 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service