Skip to Content
Merck
All Photos(1)

Key Documents

W347809

Sigma-Aldrich

1-Butanethiol

≥98%

Synonym(s):

Butyl mercaptan, Mercaptan C4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3SH
CAS Number:
Molecular Weight:
90.19
FEMA Number:
3478
Beilstein:
1730908
EC Number:
Council of Europe no.:
526
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.010
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Kosher

Agency

meets purity specifications of JECFA

vapor density

3.1 (vs air)

vapor pressure

83 mmHg ( 37.7 °C)

Assay

≥98%

refractive index

n20/D 1.443 (lit.)

bp

98 °C (lit.)

mp

−116 °C (lit.)

density

0.842 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

sulfurous

SMILES string

CCCCS

InChI

1S/C4H10S/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

WQAQPCDUOCURKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Butanethiol is a volatile sulfur-containing odorous compound that is reported to occur in skunk oil.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The history of skunk defensive secretion research.
Wood WF
The Chemical Educator, 4(2), 44-50 (1999)
Nicole W G Smits et al.
Inorganic chemistry, 58(19), 13007-13019 (2019-09-25)
The structure of the copper complex of the 6-((1-butanethiol)oxy)-tris(2-pyridylmethyl)amine ligand (Cu-tmpa-O(CH2)4SH) anchored to a gold surface has been investigated. To enable covalent attachment of the complex to the gold surface, a heteromolecular self-assembled monolayer (SAM) of butanethiol and a thiol-substituted
Nicolò Dossi et al.
Lab on a chip, 12(1), 153-158 (2011-11-15)
A sensitive and fast-responding membrane-free amperometric gas sensor is described, consisting of a small filter paper foil soaked with a room temperature ionic liquid (RTIL), upon which three electrodes are screen printed with carbon ink, using a suitable mask. It
E Mather-Mihaich et al.
Comparative biochemistry and physiology. C, Comparative pharmacology and toxicology, 85(2), 427-432 (1986-01-01)
Indicators of free-radical or oxidant-mediated responses were quantified in channel catfish, Ictalurus punctatus, exposed to the organophosphorus herbicide DEF and its metabolite, n-butyl mercaptan (nBM). The activities of the antioxidant enzymes superoxide dismutase (SOD), glutathione peroxidase (GPX) and catalase did
Late acute, delayed neurotoxic and cholinergic effects of S,S,S-tributyl phosphorotrithioite (Merphos) in hens.
M B Abou-Donia et al.
Toxicology and applied pharmacology, 53(3), 439-457 (1980-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service