Skip to Content
Merck
All Photos(1)

Key Documents

W306908

Sigma-Aldrich

Tolu balsam

blend, FG

Synonym(s):

Balsam tolu, Myroxylon toluiferum l

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
3069
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
Organoleptic:
balsam
grade:
FG
Halal
Kosher
biological source:
synthetic (mixture of 79% natural and 21% nature identical components)
synthetic
food allergen:
no known allergens

biological source

synthetic (mixture of 79% natural and 21% nature identical components)
synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

contains

Shellac as additive

mp

55-60 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsam

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tolubalsam has a sweet, cinnamon-like, tenacious odor. It is used in the perfume and soap industries as a fixative to prolong scents. It is also used as an ingredient in confectionary products, cosmetics, and as an additive to tobacco .

Application

Tolubalsam is used as a fixative in perfumery and soapmaking. (3)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Constituents of commercial Tolu balsam
Wahlberg I, et al,
Acta Chemica Scandinavica, 25(9), 3285-3295 (1971)
[The substitution of a syrup of tolu balsam with a therapeutically equivalent preparation].
E Gafiţanu et al.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi, 95(3-4), 345-346 (1991-07-01)
[Proposal for the Pharmacopeia: tolu balsam fluid extract].
M Carlassare
Bollettino chimico farmaceutico, 119(12), 731-737 (1980-12-01)
Ketaki Bhate et al.
Dermatitis : contact, atopic, occupational, drug, 21(6), 317-320 (2010-12-15)
many topical products are used in the genital region. Allergic contact dermatitis (ACD) may develop from product use or due to treatment of an underlying dermatosis. our goal was to identify the frequency of ACD and identify top allergens in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service