Skip to Content
Merck
All Photos(1)

Key Documents

W271918

Sigma-Aldrich

Methyl 2-methylbutyrate

≥98%, FCC, FG

Synonym(s):

Methyl 2-methylbutanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)COOCH3
CAS Number:
Molecular Weight:
116.16
FEMA Number:
2719
Beilstein:
1720409
EC Number:
Council of Europe no.:
2085
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.483
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.393 (lit.)

bp

115 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; ethereal

SMILES string

CCC(C)C(=O)OC

InChI

1S/C6H12O2/c1-4-5(2)6(7)8-3/h5H,4H2,1-3H3

InChI key

OCWLYWIFNDCWRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 2-methylbutyrate is an important odor-active constituent of pineapple, apple, cantaloupe, cucumber and strawberries.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Evaluation of potent odorants in cucumbers (Cucumis sativus) and muskmelons (Cucumis melo) by aroma extract dilution analysis
Schieberle P, et al.
Journal of Food Science, 55(1), 193-195 (1990)
Within-season volatile and quality differences in stored fresh-cut cantaloupe cultivars.
Beaulieu JC.
Journal of Agricultural and Food Chemistry, 53(22), 8679-8687 (2005)
Stir bar sorptive extraction (SBSE)-enantio-MDGC-MS-a rapid method for the enantioselective analysis of chiral flavour compounds in strawberries.
Kreck M, et al.
European Food Research and Technology, 213(4), 389-394 (2001)
Determination of potent odorants in apple by headspace gas dilution analysis.
Komthong P, et al.
LWT--Food Science and Technology, 39(5), 472-478 (2006)
Yukiko Tokitomo et al.
Bioscience, biotechnology, and biochemistry, 69(7), 1323-1330 (2005-07-26)
By application of aroma extract dilution analysis (AEDA) to an aroma distillate prepared from fresh pineapple using solvent-assisted flavor evaporation (SAFE), 29 odor-active compounds were detected in the flavor dilution (FD) factor range of 2 to 4,096. Quantitative measurements performed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service