Skip to Content
Merck
All Photos(1)

Key Documents

W246301

Sigma-Aldrich

Ethyl isovalerate

≥98%, FCC, FG

Synonym(s):

3-methyl butanoic acid ethyl ester, Ethyl 3-methyl butyrate, Ethyl 3-methylbutanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COOC2H5
CAS Number:
Molecular Weight:
130.18
FEMA Number:
2463
Beilstein:
1744677
EC Number:
Council of Europe no.:
442
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39022323
PubChem Substance ID:
Flavis number:
9.447
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

(EU Regulation 1223/2009)
(follows IFRA guidelines)
meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor pressure

7.5 mmHg ( 20 °C)

Assay

≥98%

refractive index

n20/D 1.396 (lit.)

bp

131-133 °C (lit.)

mp

−99 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; fruity; pineapple; sweet

SMILES string

CCOC(=O)CC(C)C

InChI

1S/C7H14O2/c1-4-9-7(8)5-6(2)3/h6H,4-5H2,1-3H3

InChI key

PPXUHEORWJQRHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl isovalerate is a flavor ester that occurs naturally in fruits such as pineapple, banana etc.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical Studies of Pineapple (Ananas sativus Lindl). I. The Volatile Flavor and Odor Constituents of Pineapple1
Haagen-Smit AJ, et al.
Journal of the American Chemical Society, 10, 1646-1650 (1945)
Optimization of solid-phase microextraction analysis for studying the change of headspace flavor compounds of banana during ripening
Liu TT & Yang TS
Journal of Agricultural and Food Chemistry, 50(4), 653-657 (2002)
Pascal Fuchsmann et al.
Journal of chromatography. A, 1601, 60-70 (2019-06-11)
Headspace in-tube extraction (HS-ITEX) and solid phase microextraction (HS-SPME) sampling, followed by gas chromatography-mass spectrometry (GC-MS), are widely used to analyze volatile compounds in various food matrices. While the extraction efficiency of volatile compounds from foodstuffs is crucial for obtaining
Johannes Niebler et al.
Phytochemistry, 109, 66-75 (2014-12-04)
Frankincense has been known, traded and used throughout the ages for its exceptional aroma properties, and is still commonly used in both secular and religious settings to convey a pleasant odor. Surprisingly, the odoriferous principle(s) underlying its unique odor profile
Amélie Slegers et al.
Molecules (Basel, Switzerland), 20(6), 10980-11016 (2015-06-18)
Developed from crosses between Vitis vinifera and North American Vitis species, interspecific hybrid grape varieties are becoming economically significant in northern areas, where they are now extensively grown for wine production. However, the varietal differences between interspecific hybrids are not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service