Skip to Content
Merck
All Photos(1)

Key Documents

W269409

Sigma-Aldrich

Methyl isobutyrate

99%, FG

Synonym(s):

Methyl 2-methyl propanoate, Methyl 2-methylpropanoate, Methyl 2-methylpropionate, Methylisobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCOOCH3
CAS Number:
Molecular Weight:
102.13
FEMA Number:
2694
Beilstein:
1740720
EC Number:
Council of Europe no.:
287
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.412
NACRES:
NA.21
Organoleptic:
fruity; floral; sweet
grade:
FG
Fragrance grade
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

99%

refractive index

n20/D 1.384 (lit.)

bp

90 °C (lit.)

density

0.891 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fruity; floral; sweet

SMILES string

COC(=O)C(C)C

InChI

1S/C5H10O2/c1-4(2)5(6)7-3/h4H,1-3H3

InChI key

BHIWKHZACMWKOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • New Method Based on the Direct Analysis in Real Time Coupled with Time-of-Flight Mass Spectrometry to Investigate the Thermal Depolymerization of Poly(methyl methacrylate).: This research presents a novel analytical technique that could be relevant for studying the thermal behavior of methyl isobutyrate in various industrial applications (Al Khulaifi et al., 2023).

  • Diagnosis and prognosis of COVID-19 employing analysis of patients′ plasma and serum via LC-MS and machine learning.: While focused on COVID-19, the study utilizes analytical methods that are applicable to the detection of complex esters such as methyl isobutyrate in biological samples (de Fátima Cobre et al., 2022).


Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service