Skip to Content
Merck
All Photos(1)

Key Documents

W269204

Sigma-Aldrich

Isovaleraldehyde

≥97%, FG

Synonym(s):

3-Methylbutanal, 3-Methylbutyraldehyde, NSC 404119

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CHO
CAS Number:
Molecular Weight:
86.13
FEMA Number:
2692
Beilstein:
773692
EC Number:
Council of Europe no.:
94
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.006
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

vapor density

2.96 (vs air)

vapor pressure

30 mmHg ( 20 °C)

Assay

≥97%

autoignition temp.

464 °F

refractive index

n20/D 1.388 (lit.)

bp

90 °C (lit.)

density

0.803 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

chocolate; ethereal; fruity; peach; sour

SMILES string

[H]C(=O)CC(C)C

InChI

1S/C5H10O/c1-5(2)3-4-6/h4-5H,3H2,1-2H3

InChI key

YGHRJJRRZDOVPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isovaleraldehyde is one of the flavor compounds identified in wheat, corn-grits and tequila.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

32.9 °F - closed cup

Flash Point(C)

0.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extrusion of flavored corn grits: Structural characteristics, volatile compounds retention and sensory acceptability
Menis MEC, et al.
LWT--Food Science and Technology, 54(2), 434-439 (2013)
Characterization of tequila flavor by instrumental and sensory analysis.
Benn SM & Peppard TL.
Journal of Agricultural and Food Chemistry, 44(2), 557-566 (1996)
Wheat flavor components
Mcwilliams M & Mackey AC
Journal of Food Science, 34(6), 493-496 (1969)
P Perpète et al.
Journal of agricultural and food chemistry, 48(6), 2384-2386 (2000-07-11)
A medium containing labeled leucine-d(10) has been used to show that Saccharomyces cerevisiae was able to produce deuterated 3-methylbutanal in a cold contact fermentation. Whereas residual unreduced Strecker aldehydes bound to polyphenols were considered until now as the main defect
Taichi Kano et al.
Journal of the American Chemical Society, 127(47), 16408-16409 (2005-11-25)
A direct asymmetric Mannich reaction using a novel axially chiral amino trifluoromethanesulfonamide (S)-3 has been developed in highly anti-selective and enantioselective manners. Thus, in the presence of a catalytic amount of (S)-3, the reactions between aldehydes and the alpha-imino ester

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service