Skip to Content
Merck
All Photos(1)

Key Documents

W246808

Sigma-Aldrich

Isoeugenol

mixture of cis and trans, 99%, FG

Synonym(s):

2-Methoxy-4-propenylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CH=CHCH3)OH
CAS Number:
Molecular Weight:
164.20
FEMA Number:
2468
Beilstein:
1909602
EC Number:
Council of Europe no.:
172
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.004
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

99%

refractive index

n20/D 1.575 (lit.)

bp

132 °C/10 mmHg (lit.)

density

1.082 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

clove; woody; spicy; sweet

SMILES string

OC1=CC=C(/C=C/C)C=C1OC

InChI

1S/C10H12O2/c1-3-4-8-5-6-9(11)10(7-8)12-2/h3-7,11H,1-2H3/b4-3+

InChI key

BJIOGJUNALELMI-ONEGZZNKSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morahem Ashengroph et al.
Applied biochemistry and biotechnology, 166(1), 1-12 (2011-10-13)
To screen strains of halotolerant or halophile bacteria which are able to convert isoeugenol to vanillin, 36 different strains of bacteria isolated from the salty environments in Iran were investigated. During growth on isoeugenol, a moderately halotolerant Gram-negative coccobacil showed
Sathya N Prasad et al.
Neurotoxicology, 33(5), 1254-1264 (2012-07-31)
Acrylamide (ACR) intoxication in its monomeric form leads to neuronal damage in both experimental animals and humans. Oxidative stress is one of the principle mechanisms related to the neurotoxicity of ACR exposure. Hence, the present study aimed to recapitulate the
Gurpreet Kaur et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(8), 2689-2695 (2012-05-23)
Isoeugenol, a component of clover oil, possesses potent anti-inflammatory and antioxidant activity. In the present study, we investigated the effect on experimentally induced adjuvant arthritis in rats. Induction of arthritis in adjuvant exposed rats was confirmed by appearance of several
Michaela Kalmes et al.
Journal of toxicology and environmental health. Part A, 75(8-10), 478-491 (2012-06-13)
The phenolic derivatives eugenol and isoeugenol, which are naturally found in essential oils of different spices, are commonly used as fragrances. Recently data demonstrated that growth suppression produced by these substances occurs in keratinocytes and that the effects may be
J Krupa et al.
The journal of physical chemistry. B, 116(36), 11148-11158 (2012-07-26)
In situ photochemical transformations of monomers of 2-methoxy-4-(prop-1-enyl)phenol (isoeugenol) and 2-methoxy-4-(prop-2-enyl)phenol (eugenol) isolated in low temperature matrices were induced by tunable UV laser light, and the progress of the reactions was followed by FTIR spectroscopy. Conformer-selective E ↔ Z geometrical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service