Skip to Content
Merck
All Photos(1)

Key Documents

W270318

Sigma-Aldrich

Methyl 2-furoate

≥98%, FG

Synonym(s):

Methyl furan-2-carboxylate, Methyl pyromucate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
FEMA Number:
2703
Beilstein:
111110
EC Number:
Council of Europe no.:
358
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.002
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 110

Assay

≥98%

refractive index

n20/D 1.487 (lit.)

bp

181 °C (lit.)

density

1.179 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

berry; mushroom; fruity; sweet; tobacco

SMILES string

COC(=O)c1ccco1

InChI

1S/C6H6O3/c1-8-6(7)5-3-2-4-9-5/h2-4H,1H3

InChI key

HDJLSECJEQSPKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Methyl 2-furoate has been identified as one of the volatile flavor compounds in tequila, okra, berrycactus and black currant juice.

Application

Methyl 2-furoate is a high-value compound, which finds applications in fine chemicals, fragrance, and flavor industries.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic study of lipase-catalyzed esterification of furoic acid to methyl-2-furoate
Zhang Y, et al.
Biochemical Engineering Journal, 161, 107587-107587 (2020)
Effect of maturity stage and storage on flavor compounds and sensory description of berrycactus (Myrtillocactus geometrizans).
Alejandro Vazquez?Cruz M, et al.
Journal of Food Science, 77(4) (2012)
Oxidative esterification of furfural by Au nanoparticles supported CMK-3 mesoporous catalysts
Radhakrishnan R, et al.
Applied Catalysis A: General, 545, 33-43 (2017)
Influence of thermal treatment on black currant (Ribes nigrum L.) juice aroma.
Varming C, et al.
Journal of Agricultural and Food Chemistry, 52(25), 7628-7636 (2004)
Volatile components of okra.
Ames JM & Macleod G.
Phytochemistry, 29(4), 1201-1207 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service