Skip to Content
Merck
All Photos(1)

Key Documents

W237701

Sigma-Aldrich

Diethyl succinate

≥99%, FCC, FG

Synonym(s):

Diethyl butanedioate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOCH2CH2COOC2H5
CAS Number:
Molecular Weight:
174.19
FEMA Number:
2377
Beilstein:
907645
EC Number:
Council of Europe no.:
438
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39022839
PubChem Substance ID:
Flavis number:
9.444
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

6 (vs air)

Assay

≥99%

refractive index

n20/D 1.42 (lit.)

bp

218 °C (lit.)

mp

−20 °C (lit.)

density

1.047 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; grape; fruity; waxy; floral

SMILES string

CCOC(=O)CCC(=O)OCC

InChI

1S/C8H14O4/c1-3-11-7(9)5-6-8(10)12-4-2/h3-6H2,1-2H3

InChI key

DKMROQRQHGEIOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl succinate can be used as a flavoring agent in the food industry.
Natural occurrence: Apple, cocoa, grape, brandy, whiskey and wine.

Application


  • Impact of indigenous Oenococcus oeni and Lactiplantibacillus plantarum species co-culture on Cabernet Sauvignon wine malolactic fermentation: Kinetic parameters, color, and aroma.: This study explores the co-culture fermentation process in Cabernet Sauvignon wine, focusing on kinetic parameters, color, and aroma. Diethyl succinate was analyzed among the volatile compounds, contributing to the wine′s aroma profile. (Zhang et al., 2024).

  • Enhancement of ester biosynthesis in blueberry wines through co-fermentation via cell-cell contact between Torulaspora delbrueckii and Saccharomyces cerevisiae.: The research investigates the role of cell-cell contact in ester biosynthesis during the co-fermentation of blueberry wines. Diethyl succinate is one of the key esters produced, enhancing the aroma and flavor profile of the wine. (Wang et al., 2024).

  • Enhancing blueberry wine aroma: Insights from cultivar selection and berry sorting.: This article examines how different blueberry cultivars and sorting methods impact the aroma of blueberry wine, with a focus on the production of aromatic esters such as diethyl succinate. (Wang et al., 2023).

  • Comparative analysis of microbial communities and volatile flavor components in the brewing of Hongqu rice wines fermented with different starters.: The study provides a comparative analysis of microbial communities and their impact on volatile flavor components, including diethyl succinate, in Hongqu rice wines fermented with different starters. (Chen et al., 2023).

  • Update to RIFM fragrance ingredient safety assessment, diethyl succinate, CAS Registry Number 123-25-1.: This update focuses on the safety assessment of diethyl succinate as a fragrance ingredient, providing comprehensive toxicological data and usage guidelines. (Api et al., 2023).

Biochem/physiol Actions

Taste at 10-100 ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock, GA
Encyclopedia of Food and Color Additives, 1, 822-823 (1997)
Burdock, GA
Encyclopedia of Food and Color Additives, 1, 822-823 (1997)
Niki M Zacharias et al.
Journal of the American Chemical Society, 134(2), 934-943 (2011-12-08)
The Krebs tricarboxylic acid cycle (TCA) is central to metabolic energy production and is known to be altered in many disease states. Real-time molecular imaging of the TCA cycle in vivo will be important in understanding the metabolic basis of
M B Ashour et al.
Toxicology and applied pharmacology, 89(3), 361-369 (1987-07-01)
Treatment with 0.5% (w/w) dietary clofibrate, a peroxisome proliferator, for 14 days induced microsomal carboxylesterase activities for five substrates including malathion, clofibrate, diethylsuccinate, diethylphthalate, and p-nitrophenylacetate in liver and kidney of male Swiss-Webster mice and Sprague-Dawley rats. The induction was
M B Ashour et al.
Biochemical pharmacology, 36(12), 1869-1879 (1987-06-15)
A series of substituted trifluoroketones were tested as inhibitors of mammalian liver microsomal carboxylesterase(s) hydrolyzing a variety of substrates including malathion, diethylsuccinate (DES) and p-nitrophenyl acetate (p-NpAc). The trifluoroketones used were very potent "transition state" inhibitors of crude mouse and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service