Skip to Content
Merck
All Photos(1)

Key Documents

W225800

Sigma-Aldrich

Cassia oil

FG

Synonym(s):

Cinnamomum cassia

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2258
Council of Europe no.:
401n
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
Organoleptic:
woody; spicy
grade:
FG
Halal
Kosher
biological source:
Cinnamomum cassia Blume
food allergen:
no known allergens

biological source

Cinnamomum cassia Blume

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.20

refractive index

n20/D 1.609

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody; spicy

Related Categories

Biochem/physiol Actions

Taste at 10 ppm

Preparation Note

Extraction method: steam distillation

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

197.0 °F - closed cup

Flash Point(C)

91.67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Yi et al.
International journal of molecular sciences, 19(5) (2018-04-26)
The present study was to determine the efficacy of dietary supplementation with oleum cinnamomi (OCM) on growth performance and intestinal functions in piglets. Sixteen piglets (24-day-old) were randomly assigned to the control or OCM groups. Piglets in the control group
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 105-105 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service