Skip to Content
Merck
All Photos(3)

Key Documents

W236217

Sigma-Aldrich

Decanal

greener alternative

natural, ≥97%, FG

Synonym(s):

Caprinaldehyde, Decyl aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CHO
CAS Number:
Molecular Weight:
156.27
FEMA Number:
2362
Beilstein:
1362530
EC Number:
Council of Europe no.:
98c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.010
NACRES:
NA.21
Organoleptic:
orange; citrus; floral; waxy; sweet
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
Cocos nucifera
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
tree nuts

biological source

Cocos nucifera

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 182.60

vapor density

>1 (vs air)

vapor pressure

~0.15 mmHg ( 20 °C)

Assay

≥97%

form

liquid

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

refractive index

n20/D 1.428 (lit.)

bp

207-209 °C (lit.)

density

0.83 g/mL at 20 °C
0.83 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

tree nuts

fragrance allergen

no known allergens

greener alternative category

Organoleptic

orange; citrus; floral; waxy; sweet

SMILES string

[H]C(=O)CCCCCCCCC

InChI

1S/C10H20O/c1-2-3-4-5-6-7-8-9-10-11/h10H,2-9H2,1H3

InChI key

KSMVZQYAVGTKIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Decanal is one of the volatile flavor compounds found in orange juices and beef.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”. Click here to view its Biobased natural statement.

Application


  • (14)C-Isotope Use to Quantify Covalent Reactions between Flavor Compounds and β-Lactoglobulin.: This article details a method using C-14 isotopes to track the covalent bonding of Decanal to proteins like β-Lactoglobulin, providing a new approach to understanding flavor compound interactions, essential for food science research (Shepelev and Reineccius, 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of solid-phase microextraction analysis for headspace flavor compounds of orange juice
Jia M, et al.
Journal of Agricultural and Food Chemistry, 46(7), 2744-2747 (1998)
K Hör et al.
Journal of agricultural and food chemistry, 49(1), 21-25 (2001-02-15)
Based on (2)H/(1)H ratio measurements of commercial synthetic and "natural" references, the recently developed on-line gas chromatography pyrolysis isotope ratio mass spectrometry (HRGC-P-IRMS) technique was used to determine the delta(2)H(SMOW) values of the flavor compounds decanal, linalool, and linalyl acetate
M S Wright et al.
Toxicon : official journal of the International Society on Toxinology, 38(9), 1215-1223 (2000-03-29)
The fungi Aspergillus flavus and Aspergillus parasiticus produce a potent class of hepatocarcinogens known as aflatoxins. Corn-derived volatile compounds have been previously found to affect growth and aflatoxin production in A. flavus. In this study, the effects on A. parasiticus
R W G Van Willige et al.
Food additives and contaminants, 20(1), 84-91 (2003-01-10)
The influence of flavour absorption by low-density polyethylene (LDPE), polycarbonate (PC) and polyethylene terephthalate (PET) on taste perception of a model solution containing seven flavour compounds and orange juice in glass bottles was studied with and without pieces of the
Anolda Cetkauskaite et al.
Environmental toxicology, 19(4), 372-386 (2004-07-23)
The aim of this research was to analyze the effects and the modes of action of elemental sulfur (S(0)) in bioluminescence and respiration of Vibrio fischeri cells and the enzymes crude luciferase, pure catalase, and alcohol dehydrogenase (ADH). Metallic copper

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service