Skip to Content
Merck
All Photos(1)

Key Documents

W229210

Sigma-Aldrich

Cinnamon leaf oil

Ceylon origin, FG

Synonym(s):

Cinnamomum zeylanicum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2292
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
Organoleptic:
cinnamon; clove; spicy; woody
grade:
FG
Kosher
biological source:
Cinnamonum zeylanicum Blume
food allergen:
no known allergens

biological source

Cinnamonum zeylanicum Blume

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

optical activity

[α]20/D +0.7°, neat

origin

Ceylon origin

refractive index

n20/D 1.5330

bp

194-234 °C

density

1.041 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cinnamon; clove; spicy; woody

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cinnamon leaf oil is mainly used as a food additive and flavoring agent in the food industry due to its distinct aroma. Eugenol and cinnamaldehyde are the key volatile constituents of this oil.

Preparation Note

Extraction method: steam distillation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effect of chitosan coatings enriched with cinnamon oil on the quality of refrigerated rainbow trout
Ojagh SM, et al
Food Chemistry, 120(1), 193-198 (2010)
Historical overview of the cinnamon industry
Wijesekera RO
Critical Reviews in Food Science and Nutrition, 10(1), 1-30 (1977)
Fungicidal activity of essential oils of Cinnamomum zeylanicum (L.) and Syzygium aromaticum (L.) Merr et LM Perry against crown rot and anthracnose pathogens isolated from banana
Ranasinghe L, et al
Letters in Applied Microbiology, 35(3), 208-211 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service