Skip to Content
Merck
All Photos(1)

Key Documents

W209910

Sigma-Aldrich

Anisyl alcohol

natural, ≥98%, FG

Synonym(s):

4-Methoxybenzyl alcohol, p-Anisyl alcohol, Anis alcohol, Anise alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2OH
CAS Number:
Molecular Weight:
138.16
FEMA Number:
2099
Beilstein:
636654
EC Number:
Council of Europe no.:
66
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.128
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

composition

Contains Anise alcohol

refractive index

n20/D 1.544 (lit.)

bp

259 °C (lit.)

mp

22-25 °C (lit.)

density

1.113 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

anise alcohol

Organoleptic

anise; honey; floral; sweet; vanilla

SMILES string

COc1ccc(CO)cc1

InChI

1S/C8H10O2/c1-10-8-4-2-7(6-9)3-5-8/h2-5,9H,6H2,1H3

InChI key

MSHFRERJPWKJFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: Anise, honey, bourbon, vanilla.

Application


  • De Novo Biosynthesis of Anisyl Alcohol and Anisyl Acetate in Engineered Escherichia coli.: This study discusses the biosynthesis of anisyl alcohol and its acetate derivatives in engineered E. coli strains. It highlights the potential applications of these compounds in the food and flavor industries due to their aromatic properties. The research demonstrates the feasibility of microbial production of anisyl alcohol as a sustainable alternative to traditional chemical synthesis (Pan et al., 2023).

  • Aryl-alcohol oxidase involved in lignin degradation: a mechanistic study based on steady and pre-steady state kinetics and primary and solvent isotope effects with two alcohol substrates.: This paper investigates the role of aryl-alcohol oxidase in the degradation of lignin, focusing on the enzyme′s kinetics and mechanisms. Anisyl alcohol is used as a substrate to study these processes, providing insights into the enzyme′s function and its potential applications in biotechnological lignin valorization (Ferreira et al., 2009).

  • Biotransformations of propenylbenzenes by an Arthrobacter sp. and its t-anethole blocked mutants.: This research explores the biotransformation of propenylbenzenes, including anisyl alcohol, using an Arthrobacter species and its mutants. The study highlights the microbial conversion pathways and the potential for producing valuable aromatic compounds through biocatalysis (Shimoni et al., 2003).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 24, 49-49 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service