Skip to Content
Merck
All Photos(1)

Key Documents

W204600

Sigma-Aldrich

Almond oil, bitter

natural, FG, ≥97%

Synonym(s):

Almond essential oil, Bitter almond oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2046
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
tree nuts
Pricing and availability is not currently available.

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.20

Assay

≥97%

form

liquid

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

tree nuts

fragrance allergen

benzaldehyde

Organoleptic

almond; bitter

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

142.0 °F - closed cup

Flash Point(C)

61.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service