SYNS181125
Synthware™ straight bore PTFE stopcock plug
bore size 2 mm, plug size: 11/25
Synonym(s):
Synthware S181125
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S181125
bore size
2 mm
Related Categories
General description
This replacement kit is complete with one PTFE plug (1:5 taper), one PTFE washer, one O-ring.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)