SYNM164380
Synthware™ solvent transfer manifold
joint: ST/NS 24/40
Synonym(s):
Synthware M164380,
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS24/40
manufacturer/tradename
M164380
General description
Synthware™ solvent transfer manifold is used for purifying and transferring solvents under inert conditions. It is supplied with a high vacuum valve featuring a PTFE O-ring at the tip and a PTFE wiper to protect the Viton® O-ring on the shaft.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)