Key Documents
Z164038
Sigma-Aldrich
Aldrich® cold-finger condenser
joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint: ST/NS24/40
Overall H
310 mm
cold-finger L × diam.
250 mm × 80 mm
Related Categories
General description
For reactions using low-boiling solvents.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)