SYNF901410
Synthware™ pear-shaped Schlenk flask with stopcock
capacity 10 mL, joint: ST/NS 14/20
Synonym(s):
Synthware F901410, modified Schlenk reaction flask
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
Recommended Products
material
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F901410
capacity
10 mL
Related Categories
General description
A suffix of G indicates a glass stopcock, H indicates a high-vacuum valve, no letter suffix indicates a PTFE Stopcock.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)