SYNF671425
Synthware™ pressure equalizing addition funnel with PTFE stopcock in equalizing line
25 mL, joint: ST/NS 14/20, stopcock: PTFE
Synonym(s):
Synthware F671425, addition funnel
All Photos(1)
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
Recommended Products
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F671425
capacity
25 mL
25 mL
Related Categories
General description
With straight bore PTFE stopcock in pressure equalizing line, graduated.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)