Skip to Content
Merck
All Photos(2)

Key Documents

424838

Sigma-Aldrich

Triphenylgermanium hydride

Synonym(s):

Triphenylgermane, Triphenylgermanyl hydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3GeH
CAS Number:
Molecular Weight:
304.96
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder or crystals
solid

reaction suitability

core: germanium
reagent type: reductant

mp

40-43 °C (lit.)

SMILES string

c1ccc(cc1)[GeH](c2ccccc2)c3ccccc3

InChI

1S/C18H16Ge/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15,19H

InChI key

NXHORDQDUDIXOS-UHFFFAOYSA-N

General description

Triphenylgermanium hydride is a hydrogen donor.

Application

It may be used in the isomerization of (Z) alkene.1 TEMPO (2,2,6,6-Tetramethylpiperidinyloxy) reacts with triphenylgermanium hydride to form its corresponding germyl radical. 2

Packaging

Packaged in glass bottles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Palladium (II)-catalyzed isomerization of olefins with tributyltin hydride.
Kim IS, et al.
The Journal of Organic Chemistry, 73(14), 5424-5426 (2007)
Vanessa A Béland et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(56), 12751-12757 (2020-04-16)
The development of batteries and fuel cells has brought to light a need for carbon anode materials doped homogeneously with electrocatalytic metals. In particular, combinations of electrocatalysts in carbon have shown promising activity. A method to derive functional carbon materials
Grace Flynn et al.
Nano letters, 16(1), 374-380 (2015-12-18)
Herein, we report the formation of multisegment Si-Ge axial heterostructure nanowires in a wet chemical synthetic approach. These nanowires are grown by the liquid injection of the respective silicon and germanium precursors into the vapor phase of an organic solvent
Theresia Ahrens et al.
Dalton transactions (Cambridge, England : 2003), 45(11), 4716-4728 (2016-02-11)
The dihydrido germyl complex cis,fac-[Rh(GePh3)(H)2(PEt3)3] (2) was synthesized by an oxidative addition of HGePh3 at [Rh(H)(PEt3)3] (1). Treatment of 2 with neohexene generated the rhodium(i) germyl complex [Rh(GePh3)(PEt3)3] (3). Alternatively, treatment of the methyl complex [Rh(CH3)(PEt3)3] (4) with HGePh3 furnished
E Mullane et al.
Physical chemistry chemical physics : PCCP, 17(10), 6919-6924 (2015-02-14)
Here we describe a relatively facile synthetic protocol for the formation of Si-Ge and Si-Ge-Si1-xGex axial nanowire heterostructures. The wires are grown directly on substrates with an evaporated catalytic layer placed in the vapour zone of a high boiling point

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service