Skip to Content
Merck
All Photos(1)

Key Documents

SYND452920

Synthware Dean Stark distillation receiver

20 mL, joint: ST/NS 29/42

Synonym(s):

Synthware D452920, Dean-Stark apparatus, Dean-Stark trap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104812
NACRES:
NB.51

Joint

joint: ST/NS29/42

packaging

pkg of 1 ea

manufacturer/tradename

Synthware D452920

capacity

20 mL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For easy separation of water from reaction or distillation system. Graduated with 2 mm PTFE stopcock.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service