Key Documents
SYNA552440
Synthware distillation distribution adapter
joint: ST/NS 24/40, Distribution Adapter
Synonym(s):
Synthware A552440, cow distillation adapter
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A552440
Related Categories
General description
Rotating cow receiver allows four flasks to be progressively filled.
Other Notes
includes only the top part
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)