Key Documents
SYND240001
Synthware distillation column packing, mini coil style
O.D. 4 mm, L 12 mm, weight 500 g
Synonym(s):
Synthware D240001
About This Item
UNSPSC Code:
40161600
NACRES:
NB.51
manufacturer/tradename
D240001
L
12 mm
O.D.
4 mm
weight
500 g
Related Categories
General description
Synthware distilling column packing material is a particle-type mini-glass coil packing that can be "poured" into any laboratory or pilot-scale column. It is easily removable for replacement or cleaning.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)